missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL18R1 Monoclonal specifically detects IL18R1 in Mouse samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | IL18R1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 13 |
| Conjugate | Alexa Fluor 488 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL18R1 (NP_032391.1; Met 1-Gly 326). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?