Learn More
Invitrogen™ IL18BPd Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA547097
Description
In Western blots, approximately 25% cross-reactivity with recombinant mouse IL-18 BPc is observed and less than 2% cross-reactivity with recombinant human IL-18 BPa is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
| IL18BPd | |
| Polyclonal | |
| Unconjugated | |
| Il18bp | |
| Igifbp; Il18bp; IL-18BP; interferon gamma inducing factor binding protein; interferon gamma-inducing factor-binding protein; interleukin 18 binding protein; interleukin-18-binding protein; MC54L | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 16068 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9Z0M9 | |
| Il18bp | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-18 BPd Thr27-Ala191. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.