Learn More
Invitrogen™ IL17RE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553891
Description
Immunogen sequence: EKSHHISIPS PDISHKGLRS KRTQPSDPET WESLPRLDSQ RHGGPEFSFD LLPEARAIRV TISSGPEVSV RLCHQWALEC EELSSPYDVQ KIVSGGHTVE LPYEFLLPCL CMEASYLQED TVRRKKCPFQ SWPEAYGSDF WKSVHFTDYS Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 75%.
The interleukin 17 receptor E (IL-17RE) shares limited similarity to the receptor of IL-17A, a ubiquitous type I membrane glycoprotein that with IL-17A plays a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. IL-17RE is the functional receptor of IL-17C and has been suggested to mediate mucosal immunity to infection with intestinal pathogens. IL-17C is produced by epithelia in response to bacterial challenge and binds to a receptor complex formed by IL-17RE and IL-17RA on the epithelial cell surface, thereby regulating the immune function of epithelial cells in an autocrine manner.
Specifications
| IL17RE | |
| Polyclonal | |
| Unconjugated | |
| IL17RE | |
| AA589509; IL-17 receptor E; IL17RE; IL-17RE; Il25r; interleukin 17 receptor E; interleukin-17 receptor E; UNQ3056/PRO9877 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 132014 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFR9 | |
| IL17RE | |
| Recombinant protein corresponding to Human IL17RE. Recombinant protein control fragment (Product #RP-90384). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.