missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL17RD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111112
This item is not returnable.
View return policy
Description
Immunogen sequence: LACKPFWKPR NLNISQHGSD MQVSFDHAPH NFGFRFFYLH YKLKHEGPFK RKTCKQEQTT ETTSCLLQNV SPGDYIIELV DDTNTTRKVM Highest antigen sequence indentity to the following orthologs: Rat - 82%, Mouse - 84%.
Feedback inhibitor of fibroblast growth factor mediated Ras-MAPK signaling and ERK activation. May inhibit FGF-induced FGFR1 tyrosine phosphorylation. Regulates the nuclear ERK signaling pathway by spatially blocking nuclear translocation of activated ERK without inhibiting cytoplasmic phosphorylation of ERK. Mediates JNK activation and may be involved in apoptosis. Might have a role in the early stages of fate specification of GnRH-secreting neurons.
Specifications
| IL17RD | |
| Polyclonal | |
| Unconjugated | |
| Il17rd | |
| 2810004A10Rik; AI428510; HH18; hSef; IL-17 receptor D; IL17RD; IL-17RD; IL17Rhom; IL17RLM; interleukin 17 receptor D; interleukin-17 receptor D; Interleukin-17 receptor-like protein; mSef; RGD1565237; SEF; sef homolog; Sef-S; UNQ6115/PRO20026 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54756 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFM7 | |
| Il17rd | |
| Recombinant Protein corresonding to Human IL17RD. Recombinant protein control fragment (Product #RP-96358). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction