Learn More
IL17RB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20673-1-AP-150UL
 View more versions of this product
Product Code. 16800255
  • £349.00 / 150µL
Description
The antibody is specific to IL17RB.
Recently, a number of cytokines belonging to the interleukin (IL)-17 family have been identified. These are termed as IL-17B, IL-17C (1) and IL-17E. IL-17 is a potent proinflammatory cytokine that plays roles in a number of diseases including rheumatoid arthritis, multiple sclerosis, and promotion of tumor growth. IL-17B, C, and E like IL-17 are able to induce proinflammatory responses. However, they do not bind to the IL-17 receptor suggesting that additional IL-17R related receptor might exist. Receptor for IL-17B and IL-17E has been independently isolated and has been designated as EV127 (in mouse) and IL-17Rh1 (in human), respectively. IL-17E induces activation of NF-kB pathway and like IL-17 also induces production of IL-8. IL-17Rh1 mRNA is highly expressed in Kidney and liver and at moderate level in small intestine, colon, and brain.
Specifications
Specifications
IL17RB
Polyclonal
Unconjugated
IL17RB
CRL4, Cytokine receptor like 4, EVI27, IL 17 receptor B, IL 17 receptor homolog 1, IL 17B receptor, IL 17RB, IL 17Rh1, IL17BR, IL17RB, IL17RH1, interleukin 17 receptor B, Interleukin 17B receptor
Rabbit
Antigen Affinity Chromatography
RUO
50905, 55540
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9JIP3, Q9NRM6
Il17rb
Peptide
150 μL
Primary
Human, Mouse