missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL17RA Recombinant Rabbit Monoclonal Antibody (208), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£113.00 - £359.00
Specifications
| Antigen | IL17RA |
|---|---|
| Clone | 208 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019231
![]() |
Invitrogen™
MA552436 |
100 Tests |
£359.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019483
|
Invitrogen™
MA552437 |
25 Tests |
£113.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL17RA Recombinant Monoclonal antibody specifically detects IL17RA in Mouse samples. It is validated for Flow CytometrySpecifications
| IL17RA | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.05% BSA and 0.03% ProClin 300 | |
| AW538159; CANDF5; CD127; CD217; CDw217; hIL-17R; IL-17 receptor A; IL17R; Il17ra; IL-17RA; interleukin 17 receptor A; interleukin-17 receptor A; MGC10262; VDw217 | |
| Recombinant Mouse IL17RA Protein (Thr22-Ala158). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Il17ra |
| 208 | |
| Flow Cytometry | |
| APC | |
| Rabbit | |
| Mouse | |
| Q60943 | |
| 16172 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title