missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL17C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579473
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: mouse spleen tissue, mouse thymus tissue.
Cytokine that plays a crucial role in innate immunity of the epithelium, including to intestinal bacterial pathogens, in an autocrine manner. Stimulates the production of antibacterial peptides and proinflammatory molecules for host defense by signaling through the NF-kappa-B and MAPK pathways. Acts synergically with IL22 in inducing the expression of antibacterial peptides, including S100A8, S100A9, REG3A and REG3G. Synergy is also observed with TNF and IL1B in inducing DEFB2 from keratinocytes. Depending on the type of insult, may have both protective and pathogenic properties, either by maintaining epithelial homeostasis after an inflammatory challenge or by promoting inflammatory phenotype. Enhanced IL17C/IL17RE signaling may also lead to greater susceptibility to autoimmune diseases.
Specifications
| IL17C | |
| Polyclonal | |
| Unconjugated | |
| IL17C | |
| CX2; cytokine CX2; Il17c; IL-17C; IL-21; Interleukin; interleukin 17C; interleukin-17C; UNQ561/PRO1122 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 234836, 27189 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q8K4C5, Q9P0M4 | |
| IL17C | |
| E. coli-derived human IL17C recombinant protein (Position: H19-V197). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction