missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL13RA2 Recombinant Rabbit Monoclonal Antibody (2H25L68)
IL13RA2 Recombinant Rabbit Monoclonal Antibody (2H25L68)
Brand: Invitrogen 703684
This item is not returnable.
View return policy
Description
IL13RA2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Specifications
| IL13RA2 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q14627 | |
| IL13RA2 | |
| Protein corresponding to human IL13RA2 [aa27-aa380]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry, Western Blot | |
| 2H25L68 | |
| Unconjugated | |
| IL13RA2 | |
| cancer/testis antigen 19; CD213a2; CT19; IL-13 receptor subunit alpha-2; IL13BP; IL13R; IL-13R; IL-13R subunit alpha-2; IL13RA2; IL-13RA2; IL-13R-alpha-2; interleukin 13 binding protein; interleukin 13 receptor alpha 2; interleukin 13 receptor alpha 2 chain; interleukin 13 receptor alpha chain 2; interleukin 13 receptor subunit alpha 2; interleukin 13 receptor, alpha 2; interleukin-13 receptor subunit alpha-2; interleukin-13 receptor subunit alpha-2-like; interleukin-13-binding protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3598 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction