missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL13RA2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596045

Product Code. 16598881

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DTEIKVNPPQ DFEIVDPGYL GYLYLQWQPP LSLDHFKECT VEYELKYRNI GSETWKTIIT KNLHYKDGFD LNKGIEAKIH TLLPWQCTNG SEVQSSWAET TYWISPQGIP ETKVQDMDCV YYNWQYLLCS WKPGIGVLLD TNYNLFYWYE GLDHALQCVD YIKADGQNIG CRFPYLEASD YKDFYICVNG SSENKPIRSS YFTFQLQNIV KPLPPVYLTF TRESSCEIKL KWSIPLGPIP ARCFDYEIEI REDDTTLVTA TVENETYTLK TTNETRQLCF VVRSKVNIYC SDDGIWSEWS DKQCWEGEDL SKKTLLR; Positive Samples: THP-1, Mouse spleen, Rat thymus, Rat spleen; Cellular Location: Membrane, Single-pass type I membrane protein.

IL13RA2 (interleukin-13 receptor subunit alpha-2) is closely related to IL13RA1, a subuint of the interleukin 13 receptor complex. This protein binds as a monomer to IL13 with high affinity, but lacks cytoplasmic domain, and does not appear to function as a signal mediator.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL13RA2
Polyclonal
Unconjugated
IL13RA2
cancer/testis antigen 19; CD213a2; CT19; IL-13 receptor subunit alpha-2; IL13BP; IL13R; IL-13R; IL-13R subunit alpha-2; IL13RA2; IL-13RA2; IL-13R-alpha-2; interleukin 13 binding protein; interleukin 13 receptor alpha 2; interleukin 13 receptor alpha 2 chain; interleukin 13 receptor alpha chain 2; interleukin 13 receptor subunit alpha 2; interleukin 13 receptor, alpha 2; interleukin-13 receptor subunit alpha-2; interleukin-13 receptor subunit alpha-2-like; interleukin-13-binding protein
Rabbit
Affinity Chromatography
RUO
16165, 171060, 3598
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
1.76 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
O88786, Q14627, Q8VHK6
IL13RA2
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.