missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL13RA1 Recombinant Rabbit Monoclonal Antibody (038), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£142.00 - £283.00
Specifications
| Antigen | IL13RA1 |
|---|---|
| Clone | 038 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019565
![]() |
Invitrogen™
MA552264 |
100 Tests |
£283.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019885
|
Invitrogen™
MA552265 |
25 Tests |
£142.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL13RA1 Recombinant Monoclonal antibody specifically detects IL13RA1 in Human samples. It is validated for Flow CytometrySpecifications
| IL13RA1 | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.5% BSA and 0.03% ProClin 300 | |
| AI882074; Cancer/testis antigen 19; CD antigen CD213a1; CD213a1; CD213a1 antigen; CT19; IL13 receptor alpha-1 chain; IL-13 receptor subunit alpha-1; IL13R; IL-13R subunit alpha-1; IL-13r[a]; IL13RA; IL-13Ra; IL13RA1; IL-13RA1; IL-13R-alpha-1; interleukin 13 receptor subunit alpha 1; interleukin 13 receptor, alpha 1; interleukin 13 receptor, alpha 1-like; interleukin-13 receptor subunit alpha-1; Interleukin-13-binding protein; LOC100360218; Novel cytokine receptor 4; NR4 | |
| Recombinant Human IL-13RA1/CD213a1 protein (Gly 22-Thr343). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| IL13RA1 |
| 038 | |
| Flow Cytometry | |
| PE | |
| Rabbit | |
| Human | |
| P78552 | |
| 3597 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title