missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL13RA1 Monoclonal Antibody (07)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA530530
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IL13RA1/IL-13RA1.
The protein encoded by this gene is a subunit of the interleukin 13 receptor. This subunit forms a receptor complex with IL4 receptor alpha, a subunit shared by IL13 and IL4 receptors. This subunit serves as a primary IL13-binding subunit of the IL13 receptor, and may also be a component of IL4 receptors. This protein has been shown to bind tyrosine kinase TYK2, and thus may mediate the signaling processes that lead to the activation of JAK1, STAT3 and STAT6 induced by IL13 and IL4.
Specifications
| IL13RA1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P78552 | |
| IL13RA1 | |
| Recombinant Human IL13RA1/IL-13RA1 Protein (Met1-Thr343). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 07 | |
| Unconjugated | |
| IL13RA1 | |
| AI882074; Cancer/testis antigen 19; CD antigen CD213a1; CD213a1; CD213a1 antigen; CT19; IL13 receptor alpha-1 chain; IL-13 receptor subunit alpha-1; IL13R; IL-13R subunit alpha-1; IL-13r[a]; IL13RA; IL-13Ra; IL13RA1; IL-13RA1; IL-13R-alpha-1; interleukin 13 receptor subunit alpha 1; interleukin 13 receptor, alpha 1; interleukin 13 receptor, alpha 1-like; interleukin-13 receptor subunit alpha-1; Interleukin-13-binding protein; LOC100360218; Novel cytokine receptor 4; NR4 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3597 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction