missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL12B Antibody (OTI1A3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71032H
This item is not returnable.
View return policy
Description
IL12B Monoclonal specifically detects IL12B in Human samples. It is validated for Western Blot.
Specifications
| IL12B | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| IL12B | |
| Full length recombinant protein of human IL12B (NP_002178) produced in SF9 cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI1A3 | |
| Western Blot | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
| 3593.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction