missing translation for 'onlineSavingsMsg'
Learn More

IL12B Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

£161.00 - £383.00

Specifications

Antigen IL12B
Dilution Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30226592
View Documents
Novus Biologicals
NBP3-33402-20ul
20 μL
£161.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30229778
View Documents
Novus Biologicals
NBP3-33402-100ul
100 μL
£383.00
100µL
Ver productos alternativos
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

IL12B Monoclonal antibody specifically detects IL12B in Human samples. It is validated for ELISA,Western Blot
Especificaciones

Especificaciones

IL12B
ELISA, Western Blot
Unconjugated
Rabbit
Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
PBS (pH 7.3), 50% glycerol, 0.05% BSA
3593
IgG
Affinity purified
Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL
Monoclonal
Purified
RUO
Human
CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40
Recombinant fusion protein containing a sequence corresponding to amino acids 23-328 of human IL12B (NP_002178.2).,, Sequence:, IWELKKDVYVVELDWYPDAPGEMVVLTCDTPEEDGITWTLDQSSEVLGSGKTLTIQVKEFGDAGQYTCHKGGEVLSHSLLLLHKKEDGIWSTDILKDQKEPKNKTFLRCEAKNYSGRFTCWWLTTISTDLTFSVKSSRGSSDPQGVTCGAATLSAERVRGDNKEYEYSVECQEDSACPAAEESLPIEVMVDAVHKLKYENYTSSFFIRDIIKPDPPKNLQLKPLKNSRQVEVSWEYPDTWSTPHSYFSLTFCVQVQGKSKREKKDRVFTDKTSATVICRKNASISVRAQDRYYSSSWSEWASVPCS
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documentos

Documentos

Certificados
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado