missing translation for 'onlineSavingsMsg'
Learn More

IL12B Antibody (2H6), Novus Biologicals™

Product Code. 18387688 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387688 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387688 Supplier Novus Biologicals Supplier No. H00003593M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL12B Monoclonal antibody specifically detects IL12B in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL12B
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2H6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002178
Gene Alias CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40
Host Species Mouse
Immunogen IL12B (NP_002178, 229 a.a. ~ 328 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IRDIIKPDPPKNLQLKPLKNSRQVEVSWEYPDTWSTPHSYFSLTFCVQVQGKSKREKKDRVFTDKTSATVICRKNASISVRAQDRYYSSSWSEWASVPCS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3593
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.