missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL12B Antibody (10B10), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | IL12B |
|---|---|
| Clone | 10B10 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30024426
|
Novus Biologicals
NBP3-26203-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028286
|
Novus Biologicals
NBP3-26203-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL12B Monoclonal antibody specifically detects IL12B in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| IL12B | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40 | |
| A synthesized peptide derived from Human IL12B [UniProt P29460] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 10B10 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3593 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title