Learn More
Invitrogen™ IL11RA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA593108
Description
Immunogen sequence: WGPPGVQYGQ PGRSVKLCCP GVTAGDPVSW FRDGEPKLLQ GPDSGLGHEL VLAQADSTDE GTYICQTLDG ALGGTVTLQL GYPPARPVVS CQAADYENFS CTWSPSQISG LPTRYLTSYR KKTVLGADSQ RRSPSTGPWP CPQDPLGAAR CVVHGAEFWS QYRINVTEVN PLGASTRLLD VSLQSILRPD PPQGLRVESV PGYPRRLRAS WTYPASWPCQ PHFLLKFRLQ YRPAQHPAWS TVEPAGLEEV ITDAVAGLPH A; Positive Samples: Mouse kidney, Mouse heart, Mouse skeletal muscle, Rat liver; Cellular Location: Membrane, Single-pass type I membrane protein.
Interleukin 11 is a stromal cell-derived cytokine that belongs to a family of pleiotropic and redundant cytokines that use the gp130 transducing subunit in their high affinity receptors. This gene encodes the IL-11 receptor, which is a member of the hematopoietic cytokine receptor family. This particular receptor is very similar to ciliary neurotrophic factor, since both contain an extracellular region with a 2-domain structure composed of an immunoglobulin-like domain and a cytokine receptor-like domain. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Jun 2012].
Specifications
| IL11RA | |
| Polyclonal | |
| Unconjugated | |
| IL11RA | |
| AI314697; CRSDA; Enhancer trap locus homolog 2; Etl2; etl-2; GP130; IL-11 receptor subunit alpha; IL-11 receptor subunit alpha-1; IL-11R subunit alpha; IL-11R subunit alpha-1; Il11ra; IL-11RA; Il11ra1; IL-11RA1; Il11ra2; Il-11ra-alpha; IL-11R-alpha; IL-11R-alpha-1; interleukin 11 receptor subunit alpha; interleukin 11 receptor, alpha; interleukin 11 receptor, alpha chain 1; interleukin 11 receptor, alpha chain 2; interleukin-11 receptor alpha chain; interleukin-11 receptor subunit alpha; Interleukin-11 receptor subunit alpha-1; locus 2; novel cytokine receptor 1; NR1; NR-1; sIL11RA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16157, 245983, 3590 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.89 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14626, Q64385, Q99MF4 | |
| IL11RA, Il11ra1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-290 of human IL11RA (NP_0011362561). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.