missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL10RA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13356-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a receptor for interleukin 10. This protein is structurally related to interferon receptors. It has been shown to mediate the immunosuppressive signal of interleukin 10, and thus inhibits the synthesis of proinflammatory cytokines. This receptor is reported to promote survival of progenitor myeloid cells through the insulin receptor substrate-2/PI 3-kinase/AKT pathway. Activation of this receptor leads to tyrosine phosphorylation of JAK1 and TYK2 kinases. Two transcript variants, one protein-coding and the other not protein-coding, have been found for this gene.Specifications
| IL10RA | |
| Polyclonal | |
| Unconjugated | |
| IL10RA | |
| CDW210A, HIL 10R, IL 10 receptor subunit alpha, IL 10R subunit 1, IL 10R subunit alpha, IL 10R1, IL 10RA, IL-10 R alpha, IL10R, IL10RA, interleukin 10 receptor, alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3587 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13651 | |
| IL10RA | |
| IL10RA Fusion Protein Ag4156 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction