missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL10RA Chimeric Recombinant Rabbit Monoclonal Antibody (1B1.3a), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA548250
This item is not returnable.
View return policy
Description
Specificity: This antibody recognises mouse IL-10R which is a cell surface glycoprotein and a receprot for anti-inflammatory IL-10. The epitope recognised by this antibody has been localised to the ligand binding domain within the extracellular region of mouse IL-10R.
The protein encoded by this gene is a receptor for interleukin 10. This protein is structurally related to interferon receptors. It has been shown to mediate the immunosuppressive signal of interleukin 10, and thus inhibits the synthesis of proinflammatory cytokines. This receptor is reported to promote survival of progenitor myeloid cells through the insulin receptor substrate-2/PI 3-kinase/AKT pathway. Activation of this receptor leads to tyrosine phosphorylation of JAK1 and TYK2 kinases. Two transcript variants, one protein-coding and the other not protein-coding, have been found for this gene.Specifications
IL10RA Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
Q61727 | |
Il10ra | |
This antibody was raised by immunising rats with the purified recombinant extracellular region of mouse Interleukin 10 Receptor. | |
200 μg | |
Primary | |
Mouse | |
Liquid |
Flow Cytometry, Immunocytochemistry, Inhibition Assays | |
1B1.3a | |
Unconjugated | |
Il10ra | |
AW553859; CD210; CD210a; CDw210; CDW210A; CDW210B; CRF2-4; CRFB4; D21S58; D21S66; HIL-10R; I10R; I10R1; il-10 receptor; il-10 receptor antagonist; IL-10 receptor subunit alpha; IL10R; IL-10R alpha; IL-10R subunit 1; IL-10R subunit alpha; IL-10R1; IL-10R2; Il10ra; IL-10RA; interleukin 10 receptor subunit alpha; interleukin 10 receptor, alpha; interleukin-10 receptor alpha chain; interleukin-10 receptor subunit 1; Interleukin-10 receptor subunit alpha; mIL-10R | |
Rabbit | |
Affinity chromatography | |
RUO | |
16154 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG κ |