missing translation for 'onlineSavingsMsg'
Learn More

IL-9R Mouse anti-Human, Clone: AH9R2, Novus Biologicals™

Product Code. 18391933 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391933 0.025 mg 25µg
18392483 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18391933 Supplier Novus Biologicals Supplier No. NBP3120320.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-9R Monoclonal antibody specifically detects IL-9R in Human samples. It is validated for Immunoprecipitation, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-9R
Applications Neutralization
Classification Monoclonal
Clone AH9R2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation, Block/Neutralize
Formulation PBS
Gene Alias CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor
Host Species Mouse
Immunogen This antibody was raised by immunising DBA/2 mice with P815 mastocytoma cells transfected with human IL-9R cDNA.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3581
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgG2a Kappa
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.