missing translation for 'onlineSavingsMsg'
Learn More

IL-9R Antibody (AH9R7) - Azide and BSA Free, Novus Biologicals™

Product Code. 18069123 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18069123 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18069123 Supplier Novus Biologicals Supplier No. NBP236513

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-9R Monoclonal specifically detects IL-9R in Human samples. It is validated for Flow Cytometry, ELISA, Functional, CyTOF-ready, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-9R
Applications Flow Cytometry, ELISA, Functional Assay, Blocking Assay, CyTOF
Classification Monoclonal
Clone AH9R7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, ELISA, Functional, Blocking/Neutralizing, CyTOF-ready
Formulation PBS (approx. pH 7.4) with No Preservative
Gene Alias CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor
Gene Symbols IL9R
Host Species Mouse
Immunogen Human CD129-transfected cell line
Molecular Weight of Antigen 57 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3581
Test Specificity The mouse monoclonal antibody AH9R7 recognizes CD129 IL-9R alpha, a 57 kDa type I transmembrane glycoprotein expressed at low levels by lymphocytes, blood cell progenitors, eosinophils, mast cells, epithelial cells, muscle cells and neurons.
Reconstitution Protein A purified
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.