Learn More
PeproTech IL-9 Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
£177.00 - £1468.00
Specifications
| Antigen | IL-9 |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17855163
|
PeproTech
500-P29-50UG |
50 μg |
£177.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17885093
![]() |
PeproTech
500-P29-100UG |
100 μg |
£228.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17875523
![]() |
PeproTech
500-P29-1MG |
1 mg |
£1468.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: MQGCPTLAGI LDINFLINKM QEDPASKCHC SANVTSCLCL GIPSDNCTRP CFSERLSQMT NTTMQTRYPL IFSRVKKSVE VLKNNKCPYF SCEQPCNQTT AGNALTFLKS LLEIFQKEKM RGMRGKI. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-9. Anti-Human IL-9-specific antibody was purified by affinity chromatography employing an immobilized Human IL-9 matrix. Sandwich ELISA: To detect Human IL-9 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human IL-9 (500-P29Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-9. Western Blot: To detect Human IL-9 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human IL-9 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.Specifications
| IL-9 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P15248 | |
| 3578 | |
| E.coli-derived Recombinant Human IL-9. | |
| Antigen affinity chromatography | |
| Il9 |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40 | |
| Il9 | |
| Primary | |
| -20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.