Learn More
PeproTech IL-9 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P59BT-50UG
Description
AA Sequence of recombinant protein: MQRCSTTWGI RDTNYLIENL KDDPPSKCSC SGNVTSCLCL SVPTDDCTTP CYREGLLQLT NATQKSRLLP VFHRVKRIVE VLKNITCPSF SCEKPCNQTM AGNTMSFLKS LLGTFQKTEM QRQKSRP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-9. Anti-Murine IL-9-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Murine IL-9 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine IL-9 (500-P59) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine IL-9. Western Blot: To detect Murine IL-9 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Murine IL-9 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P59BT-1MG will be provided as 2 x 500 μg
Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.
Specifications
| IL-9 | |
| Polyclonal | |
| Biotin | |
| Il9 | |
| cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16198 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P15247 | |
| Il9 | |
| E.coli-derived Recombinant Murine IL-9. | |
| 50 μg | |
| Primary | |
| Mouse | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.