missing translation for 'onlineSavingsMsg'
Learn More

IL-9 Antibody (RM9A4), DyLight 650, Novus Biologicals™

Product Code. 18339411 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339411 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339411 Supplier Novus Biologicals Supplier No. NBP312023C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-9 Monoclonal antibody specifically detects IL-9 in Mouse samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-9
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone RM9A4
Conjugate DyLight 650
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias Cytokine P40, HP40, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40
Host Species Rabbit
Immunogen This antibody was raised by immunising rats with recombinant full-length murine IL-9.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3578
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.