missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-9 Monoclonal antibody specifically detects IL-9 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | IL-9 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MH9A4 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | Cytokine P40, HP40, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40 |
| Host Species | Rabbit |
| Immunogen | This antibody was raised by immunising C57Bl/6 mice with human IL-9 coupled to ovalbumin. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?