missing translation for 'onlineSavingsMsg'
Learn More

IL-7R alpha/CD127 Antibody (BLR177J), Novus Biologicals™

Product Code. 18362857 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362857 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362857 Supplier Novus Biologicals Supplier No. NBP314749

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-7R alpha/CD127 Monoclonal antibody specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-7R alpha/CD127
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone BLR177J
Conjugate Unconjugated
Formulation Borate Buffered Saline (BBS) pH 8.2 with, BSA Free with 0.09% Sodium Azide
Gene Alias CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Host Species Rabbit
Immunogen residues 21-236 (ECD)
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3575
Target Species Human
Content And Storage Store at 2 - 8°C/ 1 year from date of receipt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.