Learn More
PeproTech IL-7 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P310BT-50UG
Description
AA Sequence of recombinant protein: MDCHIKDKDG KAFGSVLMIS INQLDKMTGT DSDCPNNEPN FFKKHLCDDT KEAAFLNRAA RKLRQFLKMN ISEEFNDHLL RVSDGTQTLV NCTSKEEKTI KEQKKNDPCF LKRLLREIKT CWNKILKGSI. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat IL-7. Anti-Rat IL-7-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat IL-7 by sandwich ELISA (using 100 mL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat IL-7 (500-P310) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat IL-7. Western Blot: To detect Rat IL-7 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 mg/mL. When used in conjunction with compatible development reagents the detection limit for Recombinant Rat IL-7 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P310BT-1MG will be provided as 2 x 500 μg
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
Specifications
| IL-7 | |
| Polyclonal | |
| Biotin | |
| Il7 | |
| A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25647, 294797 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P56478 | |
| Il7, IL7r | |
| E.coli-derived Recombinant Rat IL-7. | |
| 50 μg | |
| Primary | |
| Rat | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.