missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-7 Antibody (CL11132) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07998JF669
This item is not returnable.
View return policy
Description
IL-7 Monoclonal antibody specifically detects IL-7 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| IL-7 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| IL-7interleukin-7, interleukin 7 | |
| This antibody was developed using a synthetic peptide derived from P13232, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cell Markers | |
| 3574 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunocytochemistry/Immunofluorescence | |
| CL11132 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction