missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-7 Antibody (7417.111), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-837
This item is not returnable.
View return policy
Description
IL-7 Monoclonal specifically detects IL-7 in Human samples. It is validated for Western Blot, ELISA, Block/Neutralize.
Specifications
| IL-7 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA 1-2 ug/ml, Blocking/Neutralizing 50-200 molar excess | |
| IL7 | |
| Recombinant full length human IL7 protein. | |
| 0.1 mg | |
| Cell Cycle and Replication, Cytokine Research, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| 3574 | |
| Human | |
| Purified |
| Western Blot, ELISA, Blocking Assay | |
| 7417.111 | |
| Unconjugated | |
| IL-7interleukin-7, interleukin 7 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The has the ability to neutralize the biological activity of recombinant human IL-7. It may also be used as a capture in a human IL-7 ELISA. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction