missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6R alpha Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33510-100ul
This item is not returnable.
View return policy
Description
IL-6R alpha Monoclonal antibody specifically detects IL-6R alpha in Mouse samples. It is validated for Flow Cytometry,ELISA,Western Blot
Specifications
| IL-6R alpha | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, Flow Cytometry 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| CD126, gp80, IL6Q, IL6R, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-357 of mouse IL-6R alpha (NP_034689.2).,, Sequence:, LVLGSCRALEVANGTVTSLPGATVTLICPGKEAAGNVTIHWVYSGSQNREWTTTGNTLVLRDVQLSDTGDYLCSLNDHLVGTVPLLVDVPPEEPKLSCFRKNPLVNAICEWRPSSTPSPTTKAVLFAKKINTTNGKSDFQVPCQYSQQLKSFSCQVEILEGDKVYHIVSLCVANSVGSKSSHNEAFHSLKMVQPDPPANLVVSAIPGRPRWLKVSWQHPETWDPSYYLLQFQLRYRPVWSKEFTVLLLPVAQYQCVIHDALRGVKHVVQVRGKEELDLGQWSEWSPEVTGTPWIAEPRTTPAGILWNPTQVSVEDSANHEDQYESSTEATSVLAPVQE | |
| 100 μL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3570 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction