missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6R alpha Antibody (17506R), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB227RAPCCY7
This item is not returnable.
View return policy
Description
IL-6R alpha Monoclonal antibody specifically detects IL-6R alpha in Human samples. It is validated for Flow Cytometry
Specifications
| IL-6R alpha | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3570 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 17506R | |
| Flow Cytometry | |
| CD126, gp80, IL6Q, IL6R, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-6 R alpha , Leu20-Asp339, Accession # P08887 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction