missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-6 Receptor Recombinant Rabbit Monoclonal Antibody (502), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£113.00 - £359.00
Specifications
| Antigen | IL-6 Receptor |
|---|---|
| Clone | 502 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019325
![]() |
Invitrogen™
MA552202 |
100 Tests |
£359.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019774
|
Invitrogen™
MA552203 |
25 Tests |
£113.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-6 Receptor Recombinant Monoclonal antibody specifically detects IL-6 Receptor in Human samples. It is validated for Flow CytometrySpecifications
| IL-6 Receptor | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.05% BSA and 0.03% ProClin 300 | |
| CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; IL6R; IL-6R; IL-6R 1; IL-6R subunit alpha; IL6R1; IL-6R-1; Il6ra; IL-6RA; IL-6R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991 | |
| Recombinant Human IL6R/CD126 Protein (Leu 20-Pro365). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| IL6R |
| 502 | |
| Flow Cytometry | |
| PE | |
| Rabbit | |
| Human | |
| P08887 | |
| 3570 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title