Learn More
Invitrogen™ Porcine IL-6 ELISA Kit
Description
The Porcine Interleukin 6 (IL-6) ELISA quantitates Porc IL-6 in porcine serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Porc IL-6. Principle of the method The Porcine IL-6 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P26893 |
| Assay Range | 40.96 to 10,000 pg/mL |
| Assay Sensitivity | 45 pg/mL |
| Calibration Range | 45 pg/mL to 10,000 pg/mL |
| Description | IL pg/mL to 6 ELISA Kit, 96 Assays, 100 uL Sample Volume, 45 pg/mL Sensitivity, NP_001239358.1, NP_999564.1 Accession Number, 100628202; 399500 Gene ID, IL6 Gene Symbol, Swine/Porcine Species, Cytokines and Receptors Protein Family, Colorimetric Detection Method |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
| Detection Method | Colorimetric |
| Product Type | ELISA |
| Protein Family | Cytokines & Receptors |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.