missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6 Mouse anti-Human, Clone: B-E4, Novus Biologicals™
Mouse Monoclonal Antibody
£319.00 - £603.00
Specifications
| Antigen | IL-6 |
|---|---|
| Clone | B-E4 |
| Concentration | 1 mg/mL |
| Dilution | ELISA |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18328896
|
Novus Biologicals
NBP3-14585-200UL |
200 μg |
£319.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18333187
|
Novus Biologicals
NBP3-14585-500UL |
500 μg |
£603.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-6 Monoclonal antibody specifically detects IL-6 in Human samples. It is validated for ELISASpecifications
| IL-6 | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience | |
| PBS. Sterile-filtered through 0.22 um. | |
| 3569 | |
| IgG2b κ | |
| Affinity purified | |
| Antibody |
| B-E4 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 | |
| Recombinant human IL-6 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title