missing translation for 'onlineSavingsMsg'
Learn More

IL-6 Mouse anti-Human, Clone: 1A3B4, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66146-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877755

  • £130.00 / 20µL
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Interleukin 6 (IL-6) is a multifunctional 26 kD protein originally discovered in the medium of RNA-stimulated fibroblastoid cells. IL-6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be important as IL-1 and TNF-a in regulating the acute phase response. IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages and endothelial cells. IL-6 acts upon a variety of cells including myeloid progenitor cells, T cells, B cells, and hepatocytes. In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes. IL-6 potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors. IL-6 plays a critical role in B-cell differentiation to plasma cells and is a potent growth factor for plasmacytoma and myeloma. IL-6 is a very useful culture supplement for the generation of a high number of antibody-producing hybridomas. Primarily produced at sites of acute and chronic inflammation, IL-6 is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of IL-6 is implicated in a wide variety of inflammation-associated disease states including diabetes mellitus and systemic juvenile rheumatoid arthritis.
Specifications

Specifications

IL-6
Monoclonal
1.8 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P05231
IL6
IL-6 Fusion Protein Ag10646
20 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
1A3B4
Unconjugated
IL6
B cell stimulatory factor 2, BSF 2, BSF2, CDF, CTL differentiation factor, HGF, HSF, Hybridoma growth factor, IFN beta 2, IFNB2, IL 6, IL6, IL-6, Interferon beta 2, Interleukin 6
Mouse
Protein A
RUO
3569
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers