missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-6 Monoclonal Antibody (6708)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523698
This item is not returnable.
View return policy
Description
In Western blots, this antibody does not cross-react with recombinant mouse (rm) IL-6, rhOSM, rhLIF, rhIL-11, rhgp130, or rhCNTF. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.
Specifications
| IL-6 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P05231, P08505 | |
| Il6 | |
| E. coli-derived recombinant human IL-6. | |
| 500 μg | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry, Neutralization, Western Blot | |
| 6708 | |
| Unconjugated | |
| Il6 | |
| B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 16193, 3569 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction