Learn More
IL-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand:  Novus Biologicals NBP2-16957
388.45 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18766773
  • £481.00 / 0.10mL
Description
IL-6 Polyclonal specifically detects IL-6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-6
Full length human IL6 Recombinant protein.
Mouse, Rat
Immunohistochemistry (Paraffin)
0.1 mL