Learn More
IL-6 Antibody (CSTRI patent anti-IL-6) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28424JF525
Product Code. 30171626
  • £328.00 / 0.10mL
Description
IL-6 Monoclonal antibody specifically detects IL-6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-6
Monoclonal
Janelia Fluor 525
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6
IL-6/ IFNb2
0.1 mL
Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience
3569
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
CSTRI patent anti-IL-6
50mM Sodium Borate
Human
Protein A purified
RUO
Primary
Human
Purified