missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6 Antibody (B-E4), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14585R
This item is not returnable.
View return policy
Description
IL-6 Monoclonal antibody specifically detects IL-6 in Human samples. It is validated for ELISA
Specifications
| IL-6 | |
| Monoclonal | |
| DyLight 550 | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 | |
| Recombinant human IL-6 | |
| 0.1 mL | |
| Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience | |
| 3569 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| ELISA | |
| B-E4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction