missing translation for 'onlineSavingsMsg'
Learn More

IL-6 Antibody (830322) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30735270 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735270 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735270 Supplier Novus Biologicals Supplier No. FAB16092MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-6 Monoclonal antibody specifically detects IL-6 in Canine samples. It is validated for Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-6
Applications Neutralization
Classification Monoclonal
Clone 830322
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6
Host Species Mouse
Immunogen E. coli-derived recombinant canine IL-6, Thr23-Met207, Accession # P41323
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3569
Target Species Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.