missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-5 Mouse ELISA Development Kit (ABTS), PeproTech®, PeproTech®
Click to view available options
Quantity:
2 x 96 Tests
Unit Size:
Pack of 2
Description
IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
Specifications
Specifications
| Accession Number | P04401 |
| Product Type | ELISA |
| Gene ID (Entrez) | 16191 |
| Gene Symbol | Il5 |
| Quantity | 2 x 96 Tests |
| Regulatory Status | RUO |
| Storage Requirements | Refer to product documentation for component specific storage temperature. |
| Target | Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction