missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IL-5 Antibody (Abgenix anti-IL-5) - IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | IL-5 |
|---|---|
| Clone | Abgenix anti-IL-5 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30044211
|
Novus Biologicals™
NBP3-28419-100ug |
100 μg |
£257.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046841
|
Novus Biologicals™
NBP3-28419-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046869
|
Novus Biologicals™
NBP3-28419-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-5 Monoclonal antibody specifically detects IL-5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| IL-5 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| B-cell differentiation factor I, EDF, Eosinophil differentiation factor, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRFB cell differentiation factor I | |
| IL-5 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| Abgenix anti-IL-5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P05113 | |
| 3567 | |
| IgG4 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title