Learn More
IL-4R Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28331-1-AP-150UL
This item is not returnable.
View return policy
Description
The IL-4R antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IL-4R. This antibody recognizes human antigen. The IL-4R antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
| IL-4R | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 3566 | |
| IgG |
| Western Blot, Immunohistochemistry, Flow Cytometry, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human |