missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4R alpha Rabbit anti-Human, Clone: R001, Novus Biologicals™
Rabbit Monoclonal Antibody
£393.00 - £522.00
Specifications
| Antigen | IL-4R alpha |
|---|---|
| Clone | R001 |
| Applications | Neutralization |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18263518
|
Novus Biologicals
NBP2-89470-200UG |
200 μg |
£393.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18291186
|
Novus Biologicals
NBP2-89470-500UG |
500 μg |
£522.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-4R alpha Monoclonal specifically detects IL-4R alpha in Human samples. It is validated for Neutralization.Specifications
| IL-4R alpha | |
| Neutralization | |
| Unconjugated | |
| Rabbit | |
| Asthma, B Cell Development and Differentiation Markers, Cancer, Immunology, Myeloid derived Suppressor Cell | |
| 0.2 um filtered solution in Histidine and Arginine buffer, 120mM NaCl, 0.02% Tween 80, pH6.0 | |
| 3566 | |
| IgG | |
| Protein A purified |
| R001 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD124 antigen, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
| Recombinant Human IL-4R alpha Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title