missing translation for 'onlineSavingsMsg'
Learn More

IL-4R alpha Antibody [DyLight 405], Novus Biologicals™

Product Code. 30733350 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733350 Supplier Novus Biologicals Supplier No. AF6844E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IL-4R alpha Polyclonal antibody specifically detects IL-4R alpha in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-4R alpha
Applications Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CD124 antigen, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-4 R alpha , Gly24-His232, Accession # P24394
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, B Cell Development and Differentiation Markers, Cancer, Immunology, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 3566
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.