Learn More
PeproTech IL-4 Polyclonal Antibody, PeproTech®, Invitrogen™

Description
AA Sequence of recombinant protein: MHGCNDSPLR EIINTLNQVT EKGTPCTEMF VPDVLTATRN TTENELICRA SRVLRKFYFP RDVPPCLKNK SGVLGELRKL CRGVSGLNSL RSCTVNESTL TTLKDFLESL KSILRGKYLQ SCTSMS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat IL-4. Anti-Rat IL-4-specific antibody was purified by affinity chromatography employing an immobilized Rat IL-4 matrix. Sandwich ELISA: To detect Rat IL-4 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Rat IL-4 (500-P94Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat IL-4. Western Blot: To detect Rat IL-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Rat IL-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Specifications
Specifications
| Antigen | IL-4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Il4 |
| Gene Accession No. | P20096 |
| Gene Alias | B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra |
| Gene Symbols | Il4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.