Learn More
IL-4 Monoclonal Antibody (11B11), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 14-7041-85
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The 11B11 antibody reacts with mouse interleukin-4 (IL-4), a 14 kD cytokine. The 11B11 antibody is a neutralizing antibody. Mouse IL-4, also known as BCDF, BCGF-1 and BSF-1, is secreted by activated Th2 and natural killer T cells, and to a lesser extent by Th1 and mast cells. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of surface MHC class II and CD23. Applications Reported: The 11B11 antibody has been reported for use in ELISA capture, ELISPOT capture, and neutralization of IL-4 bioactivity. Applications Tested: The 11B11 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse Interleukin-4 (IL-4) in combination with the biotin BVD6-24G2 (cat. 13-7042) antibody for detection and recombinant mouse IL-4 (cat. 14-8041) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 µg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 500 pg/mL - 4 pg/mL should be included in each ELISA plate. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.Specifications
IL-4 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P07750 | |
Il4 | |
Affinity chromatography | |
RUO | |
16189 | |
4° C | |
Liquid |
Immunostaining | |
11B11 | |
Unconjugated | |
Il4 | |
B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra | |
Rat | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
For Research Use Only.