Learn More
Invitrogen™ IL-4 Human Uncoated ELISA Kit
Uncoated ELISA Kit
£367.00 - £524.00
Specifications
Accession Number | P05112 |
---|---|
Assay Range | 2-200 pg/mL |
Assay Sensitivity | 2 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15561017
|
Invitrogen™
88-7046-77 |
20 x 96 Tests |
£524.00
Pack of 20 |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
15581017
|
Invitrogen™
88-7046-88 |
10 x 96 Tests |
£367.00
Pack of 10 |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
Description
The Human Interleukin-4 (IL-4) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of Human IL-4. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.Specifications
P05112 | |
2 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
Csfgm,GMCSF,Gm-CSf,MGI-IGM | |
HRP | |
Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL | |
Interleukin-4 | |
1 hr. 45 min. |
2-200 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
3565 | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Streptavidin-HRP, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86) | |
RUO | |
2°C to 8°C | |
Human | |
24 hr. 30 min. |