missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4 Antibody (11B11) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
£228.00 - £451.00
Specifications
| Antigen | IL-4 |
|---|---|
| Clone | 11B11 |
| Dilution | Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, In vivo assay, CyTOF-ready |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, In vivo Assay, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18250821
|
Novus Biologicals
NBP2-52656-0.025MG |
0.025 mg |
£228.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18251230
|
Novus Biologicals
NBP2-52656-0.2MG |
0.2 mg |
£451.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-4 Monoclonal specifically detects IL-4 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, In vivo assay, CyTOF-ready.Specifications
| IL-4 | |
| Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, In vivo assay, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| IL4 | |
| IgG | |
| Protein A or G purified | |
| Binds to a core functional region of murine IL-4, which prevents this cytokine from interacting with its cognate receptor (Yao et al., 2006 PMID: 16361318). |
| 11B11 | |
| Flow Cytometry, ELISA, Immunohistochemistry, In vivo Assay, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| 3565 | |
| BSF-1/ IL-4 purified from serum-free supernatants of cultured EL-4 cells (a C57BL/6 thymoma) induced with 4 Beta-phorbol 12 Beta-myristate 12 alpha -acetate for 48h. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title