missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-3R alpha/CD123 Antibody (talacotuzumab) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28594JF585
This item is not returnable.
View return policy
Description
IL-3R alpha/CD123 Monoclonal antibody specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-3R alpha/CD123 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 | |
| IL-3Ra/ CD123 | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Cytokine Research, Oncogenes, Phospho Specific, Protein Kinase | |
| 3563 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| talacotuzumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction