missing translation for 'onlineSavingsMsg'
Learn More

IL-3R alpha/CD123 Antibody (18N6B12), Novus Biologicals™

Mouse Monoclonal Antibody

£167.00 - £302.00

Specifications

Antigen IL-3R alpha/CD123
Clone 18N6B12
Concentration 0.5 mg/mL
Dilution Western Blot 10ug/ml, Flow Cytometry 1ug/0.5x10^6cells∼
Applications Western Blot, Flow Cytometry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18143244
View Documents
Novus Biologicals
NBP2-27225SS
0.025 mg
£167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18161124
View Documents
Novus Biologicals
NBP2-27225
0.1 mg
£302.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-3R alpha/CD123 Monoclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

IL-3R alpha/CD123
0.5 mg/mL
Western Blot, Flow Cytometry
Unconjugated
Mouse
Cytokine Research, Phospho Specific
P26951
3563
RERVYEFLSAWSTPQRFEC (aa275-293).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
18N6B12
Western Blot 10ug/ml, Flow Cytometry 1ug/0.5x10^6cells∼
Monoclonal
Purified
RUO
Human
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
IL3RA
IgG2b κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.